Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PAP10
Entrez Gene ID
816141
Gene Name
purple acid phosphatase 10
Gene Aliases
AT2G16430, ATPAP10, F16F14.7, F16F14_7, purple acid phosphatase 10
Chromosome Location
Chr.2: 7120372 - 7123079 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_127196.4 NP_179235.1 3 - 4 803 101
NM_179629.2 NP_849960.1 2 - 3 638 101
GenBank mRNA AF492662.1 - 3 - 4 496 101
AY093236.1 - 3 - 4 544 101
BT008761.1 - 3 - 4 496 101

View Full Product Details