The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001036282.2 | NP_001031359.1 | 19 - 20 | 2035 | 78 |
NM_127248.4 | NP_179287.3 | 19 - 20 | 1973 | 78 | |
GenBank mRNA | BT002041.1 | - | 19 - 20 | 1961 | 78 |
BT008804.1 | - | 19 - 20 | 1869 | 78 |