Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TRN1
Entrez Gene ID
816198
Gene Name
transportin 1
Gene Aliases
AT2G16950, ATTRN1, F12A24.13, F12A24_13, TRANSPORTIN 1, transportin 1
Chromosome Location
Chr.2: 7353773 - 7361256 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001036282.2 NP_001031359.1 24 - 25 2456 75
NM_127248.4 NP_179287.3 24 - 25 2394 75
GenBank mRNA BT002041.1 - 24 - 25 2382 75
BT008804.1 - 24 - 25 2290 75

View Full Product Details