Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LYM2
Entrez Gene ID
816217
Gene Name
lysm domain GPI-anchored protein 2 precursor
Gene Aliases
AT2G17120, CEBiP-like 1, CL-1, F6P23.25, F6P23_25, LYP1, LysM-containing receptor protein 1, lysM domain GPI-anchored protein 2 precursor, lysm domain GPI-anchored protein 2 precursor
Chromosome Location
Chr.2: 7458930 - 7460819 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_127266.4 NP_565406.1 2 - 3 1005 107
GenBank mRNA AF428464.1 - 2 - 3 822 107
AY056221.1 - 2 - 3 794 107
AY125560.1 - 2 - 3 779 107

View Full Product Details