Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GLR2
Entrez Gene ID
816231
Gene Name
glutamate receptor 2
Gene Aliases
AT2G17260, ATGLR2, ATGLR3.1, F5J6.2, F5J6_2, GLR3.1, GLUTAMATE RECEPTOR 2, glutamate receptor 2
Chromosome Location
Chr.2: 7504748 - 7508394 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001335544.1 NP_001318239.1 3 - 4 2321 67
GenBank mRNA AY495448.1 - 3 - 4 2296 67

View Full Product Details