Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GPDHC1
Entrez Gene ID
818752
Gene Name
6-phosphogluconate dehydrogenase family protein
Gene Aliases
AT2G41540, T32G6.6, T32G6_6
Chromosome Location
Chr.2: 17325846 - 17329235 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001036448.2 NP_001031525.1 6 - 7 1529 99
NM_001336924.1 NP_001325243.1 4 - 5 2223 99
NM_129717.5 NP_181685.1 6 - 7 1766 99
NM_180021.5 NP_850352.1 5 - 6 1835 99
GenBank mRNA AJ347019.1 - 6 - 7 1423 99
AY063920.1 - 5 - 6 1677 99
AY091247.1 - 4 - 5 1268 99

View Full Product Details