Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- PGAZAT
- Entrez Gene ID
-
818785
- Gene Name
- polygalacturonase ADPG2-like protein
- Gene Aliases
-
ADPG2, ARABIDOPSIS DEHISCENCE ZONE POLYGALACTURONASE 2, AT2G41850, T11A7.5, T11A7_5, polygalacturonase abscission zone A. thaliana
- Chromosome Location
-
Chr.2: 17461655 - 17464242 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_180028.3 |
NP_850359.1 |
8 - 9 |
1281 |
59 |
View Full Product Details