Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BZIP34
Entrez Gene ID
818839
Gene Name
Basic-leucine zipper (bZIP) transcription factor family protein
Gene Aliases
AT2G42380, ATBZIP34, MHK10.10, MHK10_10
Chromosome Location
Chr.2: 17646857 - 17649062 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001336969.1 NP_001325378.1 1 - 2 885 62
NM_001336970.1 NP_001325377.1 1 - 2 876 62
NM_001336971.1 NP_001325379.1 1 - 2 967 62
NM_129800.2 NP_565970.2 1 - 2 850 62
NM_180038.2 NP_850369.1 1 - 2 967 62

View Full Product Details