Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NPG1
Entrez Gene ID
818906
Gene Name
tetratricopeptide repeat (TPR)-containing protein
Gene Aliases
AT2G43040, MFL8.10, MFL8_10, no pollen germination 1
Chromosome Location
Chr.2: 17895883 - 17899556 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001337010.1 NP_001324372.1 1 - 2 359 133
NM_180051.3 NP_850382.1 2 - 3 659 133
GenBank mRNA AF474176.1 - 2 - 3 476 133
BT046199.1 - 1 - 2 176 133

View Full Product Details