Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
INT1
Entrez Gene ID
818934
Gene Name
inositol transporter 1
Gene Aliases
AT2G43330, ATINT1, INOSITOL TRANSPORTER 1, T1O24.7, inositol transporter 1
Chromosome Location
Chr.2: 18000822 - 18004195 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_180062.3 NP_850393.1 4 - 5 1162 122
GenBank mRNA AJ973175.1 - 4 - 5 849 122
AY063901.1 - 4 - 5 1130 122
AY096505.1 - 4 - 5 849 122

View Full Product Details