Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ITPK4
Entrez Gene ID
819003
Gene Name
inositol 1,3,4-trisphosphate 5/6-kinase 4
Gene Aliases
''inositol 1, 3, 4-trisphosphate 5/6-kinase 4, 4-trisphosphate 5/6-kinase 4'', AT2G43980, AtITPK4, F6E13.11, inositol 1
Chromosome Location
Chr.2: 18208253 - 18211644 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_180076.2 NP_850407.1 3 - 4 394 99
GenBank mRNA AY070449.1 - 3 - 4 386 99
AY096566.1 - 3 - 4 354 99

View Full Product Details