The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_180087.1 | NP_850418.1 | 3 - 4 | 307 | 141 |
NM_180088.3 | NP_850419.1 | 3 - 4 | 441 | 141 | |
NM_180089.2 | NP_850420.1 | 3 - 4 | 449 | 141 | |
GenBank mRNA | AF488807.1 | - | 3 - 4 | 307 | 141 |
AJ304843.1 | - | 3 - 4 | 441 | 141 | |
AJ304844.1 | - | 3 - 4 | 441 | 141 | |
BT046114.1 | - | 3 - 4 | 307 | 141 |