Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LBD18
Entrez Gene ID
819150
Gene Name
LOB domain-containing protein 18
Gene Aliases
AT2G45420, F4L23.7, LOB domain-containing protein 18
Chromosome Location
Chr.2: 18718348 - 18720729 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_180105.2 NP_850436.1 1 - 2 623 75
GenBank mRNA AB473853.1 - 1 - 2 392 75
AF447891.1 - 1 - 2 548 75
BT025716.1 - 1 - 2 370 75

View Full Product Details