Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CNGC12
Entrez Gene ID
819253
Gene Name
cyclic nucleotide-gated channel 12
Gene Aliases
AT2G46450, ATCNGC12, F11C10.14, cyclic nucleotide-gated channel 12
Chromosome Location
Chr.2: 19065536 - 19069569 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001202829.2 NP_001189758.1 7 - 8 1422 86
NM_001202830.1 NP_001189759.1 7 - 8 1357 86
NM_001337192.1 NP_001324486.1 6 - 7 1496 86
NM_180123.6 NP_850454.5 6 - 7 1606 86

View Full Product Details