Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TLP3
Entrez Gene ID
819402
Gene Name
tubby like protein 3
Gene Aliases
AT2G47900, AtTLP3, F17A22.29, tubby like protein 3
Chromosome Location
Chr.2: 19610923 - 19613325 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001036481.1 NP_001031558.1 3 - 4 588 81
NM_180150.2 NP_850481.1 2 - 3 993 81
GenBank mRNA AY045774.1 - 2 - 3 434 81
AY074273.1 - 2 - 3 888 81
AY096604.1 - 2 - 3 434 81

View Full Product Details