Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LRS1
Entrez Gene ID
819671
Gene Name
Transducin/WD40 repeat-like superfamily protein
Gene Aliases
AT3G05090, LATERAL ROOT STIMULATOR 1, T12H1.5, T12H1_5
Chromosome Location
Chr.3: 1417893 - 1423673 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001337562.1 NP_001325515.1 10 - 11 1488 76
NM_111381.3 NP_566246.1 10 - 11 1605 76
NM_180185.3 NP_850516.1 10 - 11 1603 76
GenBank mRNA AK226843.1 - 10 - 11 1563 76
AY056292.1 - 10 - 11 1513 76
AY127022.1 - 10 - 11 1556 76
AY142674.1 - 9 - 10 1204 76
BT000631.1 - 9 - 10 1204 76

View Full Product Details