Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IMPA-1
Entrez Gene ID
819857
Gene Name
importin alpha
Gene Aliases
AIMP ALPHA, AT-IMP, AT3G06720, ATKAP ALPHA, IMPA1, IMPORTIN ALPHA, IMPORTIN ALPHA ISOFORM 1, importin alpha isoform 1
Chromosome Location
Chr.3: 2120324 - 2124166 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_111552.3 NP_187328.1 1 - 2 451 57
NM_180193.3 NP_850524.1 1 - 2 431 57
GenBank mRNA AF077528.1 - 1 - 2 264 57
AY088732.1 - 1 - 2 453 57
AY094390.1 - 1 - 2 277 57
AY125529.1 - 1 - 2 216 57

View Full Product Details