Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DGD1
Entrez Gene ID
820339
Gene Name
UDP-Glycosyltransferase superfamily protein
Gene Aliases
AT3G11670, DIGALACTOSYL DIACYLGLYCEROL DEFICIENT 1, DIGALACTOSYLDIACYLGLYCEROL SYNTHASE 1
Chromosome Location
Chr.3: 3680866 - 3684871 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_111999.4 NP_187773.1 1 - 2 1070 154
NM_180230.1 NP_850561.1 1 - 2 745 154
GenBank mRNA AF149841.1 - 1 - 2 755 154
AX765832.1 - 1 - 2 694 154
AY075649.1 - 1 - 2 745 154
AY101521.1 - 1 - 2 694 154
KJ138827.1 - 1 - 2 694 154

View Full Product Details