Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CYSB
Entrez Gene ID
820428
Gene Name
cystatin B
Gene Aliases
ARABIDOPSIS THALIANA PHYTOCYSTATIN 6, AT3G12490, ATCYS6, ATCYSB, cystatin B
Chromosome Location
Chr.3: 3960401 - 3961920 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_180239.4 NP_850570.2 3 - 4 607 89

View Full Product Details