Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DXPS1
Entrez Gene ID
821704
Gene Name
1-deoxy-D-xylulose 5-phosphate synthase 1
Gene Aliases
1-deoxy-D-xylulose 5-phosphate (DXP) synthase 2, 1-deoxy-D-xylulose 5-phosphate synthase 1, AT3G21500, DXL1, DXS-like 1, DXS2
Chromosome Location
Chr.3: 7573560 - 7576721 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001338537.1 NP_001327775.1 1 - 2 256 154
NM_113045.3 NP_566686.2 1 - 2 158 154
NM_180289.4 NP_850620.2 1 - 2 256 154

View Full Product Details