Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ARV1
Entrez Gene ID
839569
Gene Name
ARV1 family protein
Gene Aliases
AT1G01020, T25K16.2, T25K16_2
Chromosome Location
Chr.1: 6788 - 9130 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035846.2 NP_001030923.1 1 - 2 169 73
NM_001331239.1 NP_001318899.1 1 - 2 562 73
NM_001331240.1 NP_001321775.1 1 - 2 562 73
GenBank mRNA AY758070.1 - 1 - 2 169 73

View Full Product Details