Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IQD18
Entrez Gene ID
839394
Gene Name
IQ-domain 18
Gene Aliases
AT1G01110, IQ-domain 18, T25K16.10, T25K16_10
Chromosome Location
Chr.1: 51953 - 54737 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001123732.2 NP_001117204.1 4 - 5 1083 86
NM_001331250.1 NP_001318900.1 3 - 4 1062 86

View Full Product Details