Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
WRKY13
Entrez Gene ID
830096
Gene Name
WRKY DNA-binding protein 13
Gene Aliases
ARABIDOPSIS THALIANA WRKY DNA-BINDING PROTEIN 13, AT4G39410, ATWRKY13, F23K16.40, F23K16_40, WRKY DNA-binding protein 13
Chromosome Location
Chr.4: 18332606 - 18334893 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_120101.3 NP_195651.1 1 - 2 486 83
GenBank mRNA AB493729.1 - 1 - 2 382 83
AF421153.1 - 1 - 2 382 83

View Full Product Details