Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ABCD1
Entrez Gene ID
830144
Gene Name
peroxisomal ABC transporter 1
Gene Aliases
ACN2, AT4G39850, ATP-binding cassette D1, Arabidopsis thaliana ATP-binding cassette D1, AtABCD1, COMATOSE, CTS, PED3, PEROXISOME DEFECTIVE 3, PXA1, T5J17.20, T5J17_20, acetate non-utilizing 2, peroxisomal ABC transporter 1
Chromosome Location
Chr.4: 18488434 - 18497266 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001204043.2 NP_001190972.1 5 - 6 1193 120
NM_001204044.1 NP_001190973.1 5 - 6 1190 120
NM_001342558.1 NP_001328232.1 4 - 5 1557 120
NM_120148.3 NP_568072.1 5 - 6 1180 120
GenBank mRNA AF378120.1 - 5 - 6 1109 120

View Full Product Details