Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ABCD1
Entrez Gene ID
830144
Gene Name
peroxisomal ABC transporter 1
Gene Aliases
ACN2, AT4G39850, ATP-binding cassette D1, Arabidopsis thaliana ATP-binding cassette D1, AtABCD1, COMATOSE, CTS, PED3, PEROXISOME DEFECTIVE 3, PXA1, T5J17.20, T5J17_20, acetate non-utilizing 2, peroxisomal ABC transporter 1
Chromosome Location
Chr.4: 18488434 - 18497266 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001204043.2 NP_001190972.1 9 - 10 2095 130
NM_001204044.1 NP_001190973.1 9 - 10 2092 130
NM_001342558.1 NP_001328232.1 8 - 9 2459 130
NM_120148.3 NP_568072.1 9 - 10 2082 130
GenBank mRNA AF378120.1 - 9 - 10 2011 130

View Full Product Details