Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RABH1c
Entrez Gene ID
830148
Gene Name
RAB GTPase homolog H1C
Gene Aliases
AT4G39890, AtRABH1c, RAB GTPase homolog H1C, T5J17.60, T5J17_60
Chromosome Location
Chr.4: 18505919 - 18507703 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_120152.4 NP_195699.1 2 - 3 385 148
GenBank mRNA AF325109.1 - 2 - 3 192 148
AF386952.1 - 2 - 3 335 148
BT000043.1 - 2 - 3 192 148

View Full Product Details