Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- ESP4
- Entrez Gene ID
-
831597
- Gene Name
- HEAT repeat-containing protein
- Gene Aliases
-
AT5G01400, ENHANCED SILENCING PHENOTYPE 4, T10O8.110, T10O8_110
- Chromosome Location
-
Chr.5: 162522 - 171302 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_120218.2 |
NP_195760.1 |
6 - 7 |
869 |
126 |
View Full Product Details