Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- CHX26
- Entrez Gene ID
-
831698
- Gene Name
- cation/H+ exchanger 26
- Gene Aliases
-
ARABIDOPSIS THALIANA CATION/HYDROGEN EXCHANGER 26, AT5G01680, ATCHX26, F7A7.200, F7A7_200, cation/H+ exchanger 26
- Chromosome Location
-
Chr.5: 253996 - 256640 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_120246.2 |
NP_195788.2 |
1 - 2 |
216 |
64 |
View Full Product Details