Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RAPTOR2
Entrez Gene ID
831681
Gene Name
regulatory-associated protein of TOR 2 (RAPTOR2)
Gene Aliases
AT5G01770, ATRAPTOR1A, RAPTOR1A, T20L15.40, T20L15_40
Chromosome Location
Chr.5: 293675 - 302015 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001342617.1 NP_001330425.1 12 - 13 1814 85
NM_001342618.1 NP_001330424.1 12 - 13 1814 85
NM_001342619.1 NP_001330427.1 12 - 13 1783 85
NM_001342620.1 NP_001330426.1 12 - 13 1774 85
NM_120255.3 NP_195797.3 12 - 13 1774 85

View Full Product Details