Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CYCD7;1
Entrez Gene ID
831830
Gene Name
CYCLIN D7;1
Gene Aliases
AT5G02110, CYCD7, CYCLIN D7;1, T7H20.160, T7H20_160
Chromosome Location
Chr.5: 416917 - 418666 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001342646.1 NP_001332391.1 2 - 3 632 112
NM_120289.3 NP_195831.1 3 - 4 804 112

View Full Product Details