Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PRT6
Entrez Gene ID
830916
Gene Name
proteolysis 6
Gene Aliases
AT5G02300, AT5G02310, T1E22.70, T1E22_70, proteolysis 6
Chromosome Location
Chr.5: 473868 - 482942 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001342651.1 NP_001318457.1 14 - 15 6071 77
NM_001342652.1 NP_001331954.1 14 - 15 6094 77
NM_120309.2 NP_195851.2 13 - 14 5736 77

View Full Product Details