Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PRR7
Entrez Gene ID
831793
Gene Name
pseudo-response regulator 7
Gene Aliases
APRR7, AT5G02810, F9G14.120, F9G14_120, pseudo-response regulator 7
Chromosome Location
Chr.5: 637681 - 642030 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001342671.1 NP_001330923.1 6 - 7 1840 81
NM_120359.3 NP_568107.1 7 - 8 1703 81
GenBank mRNA AB046954.1 - 6 - 7 1393 81
AY039943.1 - 7 - 8 1653 81
AY142560.1 - 6 - 7 1393 81

View Full Product Details