Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
EXO70A1
Entrez Gene ID
831809
Gene Name
exocyst subunit exo70 family protein A1
Gene Aliases
AT5G03540, ATEXO70A1, F12E4.330, F12E4_330, exocyst subunit exo70 family protein A1
Chromosome Location
Chr.5: 889385 - 894205 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001125690.1 NP_001119162.1 7 - 8 1610 93
NM_001203287.1 NP_001190216.1 9 - 10 1568 93
NM_120434.5 NP_195974.2 8 - 9 1671 93
GenBank mRNA AY072155.1 - 8 - 9 1491 93
AY133751.1 - 8 - 9 1450 93

View Full Product Details