Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HSFA3
Entrez Gene ID
831749
Gene Name
heat shock transcription factor A3
Gene Aliases
ARABIDOPSIS THALIANA HEAT SHOCK TRANSCRIPTION FACTOR A3, AT-HSFA3, AT5G03720, F17C15.140, F17C15_140, heat shock transcription factor A3
Chromosome Location
Chr.5: 971142 - 974326 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001342728.1 NP_001318473.1 1 - 2 989 78
NM_120453.4 NP_195992.2 1 - 2 841 78

View Full Product Details