Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SCY1
Entrez Gene ID
816386
Gene Name
SECY homolog 1
Gene Aliases
AT2G18710, SECY homolog 1
Chromosome Location
Chr.2: 8112084 - 8114485 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_127427.3 NP_179461.1 3 - 4 1076 77
GenBank mRNA AF424550.1 - 3 - 4 1061 77
AY058103.1 - 3 - 4 1061 77
AY065093.1 - 3 - 4 1068 77
BT002632.1 - 3 - 4 1043 77

View Full Product Details