Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TAG1
Entrez Gene ID
816464
Gene Name
membrane bound O-acyl transferase (MBOAT) family protein
Gene Aliases
ABX45, AS11, AT2G19450, ATDGAT, Arabidopsis thaliana acyl-CoA:diacylglycerol acyltransferase, AtDGAT1, DGAT1, DIACYLGLYCEROL ACYLTRANSFERASE, F27F23.24, RDS1, TRIACYLGLYCEROL BIOSYNTHESIS DEFECT 1, acyl-CoA:diacylglycerol acyltransferase 1
Chromosome Location
Chr.2: 8426117 - 8429853 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_127503.3 NP_179535.1 13 - 14 1666 70
GenBank mRNA AJ131831.1 - 13 - 14 1577 70
AJ238008.1 - 13 - 14 1485 70
AY054480.1 - 13 - 14 1508 70
BT008883.1 - 13 - 14 1347 70

View Full Product Details