Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ckl5
Entrez Gene ID
816466
Gene Name
casein kinase I-like 5
Gene Aliases
AT2G19470, F3P11.7, F3P11_7, casein kinase I-like 5
Chromosome Location
Chr.2: 8433542 - 8436602 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_127505.5 NP_179537.1 4 - 5 649 141
GenBank mRNA AY099837.1 - 4 - 5 540 141
AY943844.1 - 4 - 5 342 141
BT006580.1 - 4 - 5 342 141

View Full Product Details