Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FVE
Entrez Gene ID
816471
Gene Name
Transducin family protein / WD-40 repeat family protein
Gene Aliases
ACG1, AT2G19520, ATMSI4, F3P11.12, F3P11_12, MSI4, MULTICOPY SUPPRESSOR OF IRA1 4, NFC04, NFC4, NUCLEOSOME/CHROMATIN ASSEMBLY FACTOR GROUP C, WD-40 REPEAT PROTEIN MSI4
Chromosome Location
Chr.2: 8455756 - 8459642 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_127510.5 NP_565456.2 2 - 3 599 88
GenBank mRNA AF498102.1 - 2 - 3 420 88
AY057655.1 - 2 - 3 397 88
AY059799.1 - 2 - 3 373 88
AY081447.1 - 2 - 3 349 88

View Full Product Details