Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PRF5
Entrez Gene ID
816496
Gene Name
profilin 5
Gene Aliases
AT2G19770, F6F22.20, F6F22_20, profilin 5
Chromosome Location
Chr.2: 8519485 - 8521275 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_127535.3 NP_179567.1 2 - 3 430 66
GenBank mRNA AY085645.1 - 2 - 3 347 66
DQ446526.1 - 2 - 3 274 66

View Full Product Details