The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001335692.1 | NP_001325324.1 | 1 - 2 | 70 | 75 |
NM_127610.1 | NP_565473.1 | 1 - 2 | 70 | 75 |