Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PAP20
Entrez Gene ID
824444
Gene Name
Purple acid phosphatases superfamily protein
Gene Aliases
AT3G52780, ATPAP20
Chromosome Location
Chr.3: 19561260 - 19564350 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_115138.3 NP_190846.1 5 - 6 1251 120
NM_180355.1 NP_850686.1 5 - 6 1098 120
GenBank mRNA AF492666.1 - 5 - 6 1096 120
BT029748.1 - 5 - 6 1096 120

View Full Product Details