Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
UTR6
Entrez Gene ID
825105
Gene Name
UDP-galactose transporter 6
Gene Aliases
AT3G59360, ATUTR6, UDP-GALACTOSE TRANSPORTER 6, UDP-galactose transporter 6
Chromosome Location
Chr.3: 21935278 - 21940365 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_115798.3 NP_567083.1 13 - 14 1494 97
NM_180390.3 NP_850721.1 13 - 14 1615 97
GenBank mRNA AF360241.1 - 13 - 14 1480 97
AY040039.1 - 12 - 13 1166 97

View Full Product Details