Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- UTR6
- Entrez Gene ID
-
825105
- Gene Name
- UDP-galactose transporter 6
- Gene Aliases
-
AT3G59360, ATUTR6, UDP-GALACTOSE TRANSPORTER 6, UDP-galactose transporter 6
- Chromosome Location
-
Chr.3: 21935278 - 21940365 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_180390.3 |
NP_850721.1 |
14 - 15 |
1682 |
128 |
View Full Product Details