Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DPB
Entrez Gene ID
831847
Gene Name
Transcription factor DP
Gene Aliases
AT5G03415, ATDPB, DPB TRANSCRIPTION FACTOR
Chromosome Location
Chr.5: 842289 - 845454 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001203285.1 NP_001190214.1 4 - 5 818 68
NM_180426.3 NP_850757.1 4 - 5 637 68
GenBank mRNA AB493733.1 - 3 - 4 377 68
AJ294532.1 - 3 - 4 377 68
AK228148.1 - 4 - 5 618 68

View Full Product Details