Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GLU1
Entrez Gene ID
830292
Gene Name
glutamate synthase 1
Gene Aliases
AT5G04140, F21E1.60, F21E1_60, FD-GOGAT, FERREDOXIN-DEPENDENT GLUTAMATE SYNTHASE, FERREDOXIN-DEPENDENT GLUTAMATE SYNTHASE 1, GLS1, GLUS, GLUTAMATE SYNTHASE, GLUTAMATE SYNTHASE 1, glutamate synthase 1
Chromosome Location
Chr.5: 1129785 - 1138488 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_120496.3 NP_568134.1 4 - 5 1404 69
NM_180432.2 NP_850763.1 3 - 4 1236 69

View Full Product Details