Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BTR1L
Entrez Gene ID
830322
Gene Name
binding to TOMV RNA 1L (long form)
Gene Aliases
AT5G04430, BINDING TO TOMV RNA 1, BINDING TO TOMV RNA 1S (SHORT FORM), BTR1, BTR1S, T19N18.7, binding to TOMV RNA 1L (long form)
Chromosome Location
Chr.5: 1250248 - 1253953 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_120525.4 NP_196063.1 8 - 9 1130 91
NM_180433.3 NP_850764.1 8 - 9 1193 91
GenBank mRNA AF375411.1 - 8 - 9 992 91
AY086553.1 - 8 - 9 1010 91
BT000854.1 - 7 - 8 841 91

View Full Product Details