Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SYN1
Entrez Gene ID
830432
Gene Name
Rad21/Rec8-like family protein
Gene Aliases
AT5G05490, ATREC8, AtREC8, BP2, DETERMINATE, DIF1, INFERTILE 1, MOP10.3, MOP10_3, REC8, SYNAPTIC 1
Chromosome Location
Chr.5: 1624206 - 1629269 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_120631.2 NP_196168.1 2 - 3 276 81
NM_180442.2 NP_850773.1 3 - 4 276 81
GenBank mRNA AF080619.1 - 2 - 3 279 81
AF080620.1 - 3 - 4 276 81
AJ238306.1 - 2 - 3 183 81

View Full Product Details