Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
YUC4
Entrez Gene ID
831003
Gene Name
Flavin-binding monooxygenase family protein
Gene Aliases
AT5G11320, AtYUC4, F2I11.210, F2I11_210, YUCCA4
Chromosome Location
Chr.5: 3611247 - 3613640 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_121170.3 NP_196693.1 1 - 2 896 148
NM_180477.2 NP_850808.1 1 - 2 617 148

View Full Product Details