Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MSL10
Entrez Gene ID
831081
Gene Name
mechanosensitive channel of small conductance-like 10
Gene Aliases
AT5G12080, ATMSL10, F14F18.230, F14F18_230, mechanosensitive channel of small conductance-like 10
Chromosome Location
Chr.5: 3897893 - 3901790 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001125740.2 NP_001119212.1 5 - 6 1959 115
NM_121246.5 NP_196769.1 5 - 6 2019 115
NM_180479.2 NP_850810.1 4 - 5 2132 115
GenBank mRNA AK318910.1 - 5 - 6 1893 115
AY075640.1 - 5 - 6 1942 115
BT002236.1 - 3 - 4 1759 115

View Full Product Details