Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CRN
Entrez Gene ID
831170
Gene Name
Protein kinase superfamily protein
Gene Aliases
AT5G13290, CORYNE, SOL2, SUPPRESSOR OF LLP1 2, T31B5.110, T31B5_110
Chromosome Location
Chr.5: 4252842 - 4255378 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001085108.1 NP_001078577.1 2 - 3 1040 86

View Full Product Details